Learn More
phosphate cytidylyltransferase 1, choline, beta, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PCYT1B.
Brand: Abnova H00009468-A01.50uL
Description
The protein encoded by this gene belongs to the cytidylyltransferase family. It is involved in the regulation of phosphatidylcholine biosynthesis. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: NEKRYRFQNQVDKMKEKVKNVEERSKEFVNRVEEKSHDLIQKWEEKSREFIGNFLELFGPDGAWKQMFQERSSRMLQALSpecifications
| phosphate cytidylyltransferase 1, choline, beta | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PCYT1B. | |
| PCYT1B | |
| CCT-beta/CTB | |
| Mouse | |
| 50 μL | |
| Yes | |
| 9468 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004845 | |
| PCYT1B | |
| PCYT1B (NP_004836, 236 a.a. ∼ 314 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.