Learn More
PCTAIRE protein kinase 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PCTK2.
Brand: Abnova H00005128-A01.50uL
Description
The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It has similarity to rat protein which is thought to play a role in terminally differentiated neurons. [provided by RefSeq
Sequence: NFPKYKPQPLINHAPRLDSEGIELITKFLQYESKKRVSAEEAMKHVYFRSLGPRIHALPESVSIFSLKEIQLQKDPGFRNSSYPETGHGKNRRQSMLFSpecifications
| PCTAIRE protein kinase 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PCTK2. | |
| PCTK2 | |
| PCTAIRE2 | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002595 | |
| PCTK2 | |
| PCTK2 (NP_002586, 426 a.a. ∼ 523 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 5128 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.