Learn More
PCNA Mouse, Unlabeled, Clone: 24, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610665
Description
Progression of the mammalian cell cycle is regulated in two different ways: 1) phosphorylation and dephosphorylation of key proteins, 2) synthesis and degradation of regulatory factors. The Proliferating Cell Nuclear Antigen (PCNA) was initially identified as a nuclear antigen in proliferating cells and was subsequently described as a subunit for DNA polymerase δ. Human PCNA is 262 amino acids with an apparent molecular weight of 36kDa. PCNA protein levels peak during the S-phase of the cell cycle, at which time it forms a complex with the p21 inhibitor. PCNA is almost undetectable in other phases of the cycle. Because of its unique expression, PCNA has been extensively used in studies associating the prognosis of tumor progression and neoplastic proliferation.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| PCNA | |
| Monoclonal | |
| 250μg/mL | |
| Proliferating Cell Nuclear Antigen | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 24 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.