Learn More
protocadherin beta 1, Mouse, Clone: 1H4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PCDHB1.
Brand: Abnova H00029930-M01A.200uL
Description
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]
Sequence: QFSRLVYRAQVSENSPNGSLVATVTAVDLDEGTNKAITYSLAQNPEAILKTFQIDPQNGEVRLRGPLDFEAIETYDIDIQATDGGGLSAHSKVLVEVVDVSpecifications
| protocadherin beta 1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_013340 | |
| PCDHB1 | |
| PCDHB1 (NP_037472, 241 a.a. ∼ 340 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 29930 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1H4 | |
| Mouse monoclonal antibody raised against a partial recombinant PCDHB1. | |
| PCDHB1 | |
| MGC138301/MGC138303/PCDH-BETA1 | |
| Mouse | |
| 200 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.