Learn More
protocadherin alpha 5, Mouse, Clone: 1E8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PCDHA5.
Brand: Abnova H00056143-M01A.200uL
Description
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. [provided by RefSeq
Sequence: TNEEETNFLELVLRKSLDREETQEHRLLVIATDGGKPELTGTVQLLINVLDANDNAPEFDKSIYNVRLLENAPSGTLVIKLNASDADEGINKEIVYFFSNLVLDDVKSpecifications
| protocadherin alpha 5 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_018908 | |
| PCDHA5 | |
| PCDHA5 (NP_061731, 183 a.a. ∼ 289 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 56143 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1E8 | |
| Mouse monoclonal antibody raised against a partial recombinant PCDHA5. | |
| PCDHA5 | |
| CNR6/CNRN6/CNRS6/CRNR6/PCDH-ALPHA5 | |
| Mouse | |
| 200 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.