Learn More
propionyl Coenzyme A carboxylase, alpha polypeptide, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PCCA protein.
Brand: Abnova H00005095-B01P.50ug
Description
The protein encoded by this gene is the alpha subunit of the heterodimeric mitochondrial enzyme Propionyl-CoA carboxylase. PCCA encodes the biotin-binding region of this enzyme. Mutations in either PCCA or PCCB (encoding the beta subunit) lead to an enzyme deficiency resulting in propionic acidemia. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MLSAALRTLKHVLYYSRQCLMVSRNLGSVGYDPNEKTFDKILVANRGEIACRVIRTCKKMGIKTVAIHSDVDASSVHVKMADEAVCVGPAPTSKSYLNMDAIMEAIKKTRAQAVHPGYGFLSENKEFARCLAAEDVVFIGPDTHAIQAMGDKIESKLLAKKAEVNTIPGFDGVVKDAEEAVRIAREIGYPVMIKASAGGGGKGMRIAWDDEETRDGFRLSSQEAASSFGDDRLLIEKFIDNPRHIEIQVLGDKHGNALWLNERECSIQRRNQKVVEEAPSIFLDAETRRAMGEQAVALARAVKYSSAGTVEFLVDSKKNFYFLEMNTRLQVEHPVTECITGLDLVQEMIRVAKGYPLRHKQADIRINGWAVECRVYAEDPYKSFGLPSIGRLSQYQEPLHLPGVRVDSGIQPGSDISIYYDPMISKLITYGSDRTEALKRMADALDNYVIRGVTHNIALLREVIINSRFVKGDISTKFLSDVYPDGFKGHMLTKSEKNQLLAIASSLFVAFQLRAQHFQENSRMPVIKPDIANWELSVKLHDKVHTVVASNNGSVFSVEVDGSKLNVTSTWNLASPLLSVSVDGTQRTVQCLSREAGGNMSIQFLGTVYKVNILTRLAAELNKFMLEKVTEDTSSVLRSPMPGVVVAVSVKPGDAVAEGQEICVIEAMKMQNSMTAGKTGTVKSVHCQAGDTVGEGDLLVELESpecifications
| propionyl Coenzyme A carboxylase, alpha polypeptide | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human PCCA protein. | |
| PCCA | |
| PCCA | |
| PCCA (AAH00140.1, 1 a.a. ∼ 703 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000140.2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 5095 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.