missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PCBD2 protein.
Brand: Abnova H00084105-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MAAVLGALGATRRLLAALRGQSLGLAAMSSGTHRLTAEERNQAILDLKAAGWSELSERDAIYKEFSFHNFNQAFGFMSRVALQAEKMNHHPEWFNVYNVQITLTSHDCGELTKKDVKLAKFIEKAAASVSpecifications
| pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human PCBD2 protein. | |
| PCBD2 | |
| DCOH2/DCOHM/PHS2 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_032151.3 | |
| PCBD2 | |
| PCBD2 (NP_115527.2, 1 a.a. ∼ 129 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 84105 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction