missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pre-B-cell leukemia homeobox 3, Mouse, Clone: 1A11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PBX3.
Brand: Abnova H00005090-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: FNLPNSGDMFMNMQSLNGDSYQGSQVGANVQSQVDTLRHVINQTGGYSDGLGGNSLYSPHNLNANGGWQDATTPSSVTSPTEGPGSVHSDTSNSpecifications
| pre-B-cell leukemia homeobox 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006195 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 5090 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 1A11 | |
| Mouse monoclonal antibody raised against a partial recombinant PBX3. | |
| PBX3 | |
| PBX3 | |
| PBX3 (NP_006186, 342 a.a. ∼ 434 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction