Learn More
PDZ binding kinase, Mouse, Clone: 3A11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PBK.
Brand: Abnova H00055872-M11.100ug
Description
This genes encodes a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase (MAPKK) family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis. [provided by RefSeq
Sequence: MEGISNFKTPSKLSEKKKSVLCSTPTINIPASPFMQKLGFGTGVNVYLMKRSPRGLSHSPWAVKKINPICNDHYRSVYQKRLMDEAKILKSLHHPNIVGYRAFTEANDGSSpecifications
| PDZ binding kinase | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC015191 | |
| PBK | |
| PBK (AAH15191, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 3A11 | |
| Mouse monoclonal antibody raised against a partial recombinant PBK. | |
| PBK | |
| FLJ14385/Nori-3/SPK/TOPK | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 55872 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.