Learn More
PAX8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1112
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Western blot analysis of PA1-112 detects a prominent ∽48 kDa protein in human adenocarcinoma SKOV-3 and SV40 transformed African green monkey (Cercopithecus aethiops) kidney cells. In SKOV-3 cells, 2 additional unknown bands at (∽20 kD) and at ∽150 kDa are also detected. Specificity of the antibody was confirmed in HeLa and 293T cells overexpressing full lenght PAX8. The 48 kDa band is enriched in immunoprecipitations from Cos7 lysate, however additional unknown high molecular weight bands are also enriched and detected by PA1-112.
PAX8 Polyclonal antibody specifically detects PAX8 in Human, Non-human Primate samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
PAX8 | |
Polyclonal | |
Unconjugated | |
PAX8 | |
HGNC:8622; paired box 8; paired box gene 8; paired box protein Pax-8; paired domain gene 8; PAX8; Pax-8; Pax-8 protein | |
Rabbit | |
Protein G | |
RUO | |
7849 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q06710 | |
PAX8 | |
Purified internal fragment of human recombinant PAX8 expressed in E. coli | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |