Learn More
Pax-6 Mouse anti-Human, PerCP-Cy5.5, Clone: O18-1330, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562388
Description
Pax-6 is a member of the paired box (pax) gene family whose protein products are transcription factors involved in development. Pax family members share a highly conserved DNA binding domain that contains six alpha helices (paired domain) and a homeo box domain. Pax-6 has important roles in the development of the eye, nose, central nervous system, and pancreas. Defects in Pax-6 are responsible for various eye malformations including aniridia and Peters anomaly.
The O18-1330 monoclonal antibody reacts with human Pax-6. Because the Pax-6 protein sequence is highly conserved among vertebrate species, cross-reactivity with other species is possible.
Specifications
| Pax-6 | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Pax-6 aa 406-422 Peptide | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| O18-1330 | |
| Oculorhombin, Aniridia type II protein, PAX6, AN2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.