Learn More
Pax-6 Mouse anti-Human, Alexa Fluor 647, Clone: O18-1330, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562249
Description
Pax-6 is a member of the paired box (pax) gene family whose protein products are transcription factors involved in development. Pax family members share a highly conserved DNA binding domain that contains six alpha helices (paired domain) and a homeo box domain. Pax-6 has important roles in the development of the eye, nose, central nervous system, and pancreas. Defects in Pax-6 are responsible for various eye malformations including aniridia and Peters anomaly.
Host Species: Mouse
Clone: O18-1330
Isotype: IgG2a, κ
Species Reactivity: Human
Immunogen: Human Pax-6 aa 406-422 Peptide
Flow Cytometry
Specifications
| Pax-6 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human Pax-6 aa 406-422 Peptide | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| O18-1330 | |
| Oculorhombin, Aniridia type II protein, PAX6, AN2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.