Learn More
PAX5 Monoclonal Antibody (SP34), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514585
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-14585 targets Pax-5 in IHC (P) applications and shows reactivity with Human and Mouse samples. The MA5-14585 immunogen is synthetic peptide derived from the C-terminus of human Pax-5 protein.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.Specifications
PAX5 | |
Monoclonal | |
0.067 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
Q02548 | |
PAX5 | |
Synthetic peptide derived from the C-terminus of human Pax-5 protein | |
1 mL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
SP34 | |
Unconjugated | |
PAX5 | |
ALL3; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; EBB-1; KLP; paired box 5; paired box gene 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; paired box protein Pax-5; paired domain gene 5; Pax5; Pax-5; Pax5 variant 1; Pax5 variant 2; RGD1565236; transcription factor PAX 5 | |
Rabbit | |
Protein A/G | |
RUO | |
500453, 5079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |