Learn More
PAX-5 Rat, Brilliant Violet 510, Clone: 1H9, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 563191
Description
The 1H9 monoclonal antibody clone specifically binds to human and mouse Paired box protein Pax-5. Pax-5 is a ∽50 kDa protein that is also known as B-cell-specific transcription factor and B cell specific activator protein (BSAP). Pax-5 is a member of the paired box (Pax) family of transcription factors. It is expressed in pro-B, pre-B and mature B cells. Through the Groucho family of co-repressors, Pax-5 likely functions as a transcriptional repressor of non-B-lymphoid genes during the B cell commitment process. In the early stages of B cell development, Pax-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20 and maintains B cell identity. Pax-5 suppression is involved in the upregulation of Blimp-1 leading to the development of Pax-5-negative plasma cells. Pax-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. Altered forms and expression patterns of Pax-5 have been associated with some lymphoid and nonlymphoid cancers.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
| PAX-5 | |
| Monoclonal | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Recombinant Mouse Pax-5 protein containing aa 154-284 | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 1H9 | |
| Pax5, PAX5, KLP, BSAP, EBB-1; B-cell-specific transcription factor | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.