missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAS domain containing 1, Mouse, Clone: 2B10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PASD1.
Brand: Abnova H00139135-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: MKMRGEKRRDKVNPKSSQRKLNWIPSFPTYDYFNQVTLQLLDGFMITLSTDGVIICVAENISSLLGHLPAEIVGKKLLSLLPDEEKDEVYQKIILKFPLLSpecifications
| PAS domain containing 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_173493 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 139135 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2B10 | |
| Mouse monoclonal antibody raised against a partial recombinant PASD1. | |
| PASD1 | |
| PASD1 | |
| PASD1 (NP_775764, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction