missing translation for 'onlineSavingsMsg'
Learn More
Learn More
parvin, gamma, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PARVG protein.
Brand: Abnova H00064098-B01.50uL
This item is not returnable.
View return policy
Description
Members of the parvin family, including PARVG, are actin-binding proteins associated with focal contacts.[supplied by OMIM
Sequence: MEPEFLYDLLQLPKGVEPPAEEELSKGGKKKYLPPTSRKDPKFEELQKVLMEWINATLLPEHIVVRSLEEDMFDGLILHHLFQRLAALKLEAEDIALTATSQKHKLTVVLEAVNRSLQLEEWQAKWSVESIFNKDLLSTLHLLVALAKRFQPDLSLPTNVQVEVITIESTKSGLKSEKLVEQLTEYSTDKDEPPKDVFDELFKLAPEKVNAVKEAIVNFVNQKLDRLGLSVQNLDTQFADGVILLLLIGQLEGFFLHLKEFYLTPNSPAEMLHNVTLALELLKDEGLLSCPVSPEDIVNKDAKSTLRVLYGLFCKHTQKAHRDRTPHGAPNSpecifications
| parvin, gamma | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human PARVG protein. | |
| PARVG | |
| PARVG | |
| PARVG (NP_071424.1, 1 a.a. ∼ 331 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_022141.4 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 64098 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction