missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPPA Mouse anti-Human, Clone: 5H9, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA146425
This item is not returnable.
View return policy
Description
This antibody works well in htPAPP-A and dPAPP-A (PAPP-A dimer) immunoassays.
This gene encodes a secreted metalloproteinase which cleaves insulin-like growth factor binding proteins (IGFBPs). It is thought to be involved in local proliferative processes such as wound healing and bone remodeling. Low plasma level of this protein has been suggested as a biochemical marker for pregcies with aneuploid fetuses.
Specifications
| PAPPA | |
| Monoclonal | |
| 1 mg/mL | |
| PAPPA | |
| RP11-45A16.1, ASBABP2, DIPLA1, IGFBP-4ase, PAPA, PAPP-A, PAPPA1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5069 | |
| 4° C, do not freeze | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 5H9 | |
| Unconjugated | |
| Q13219 | |
| PAPPA | |
| Purified human PAPPA. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction