Learn More
Invitrogen™ HPV/BPV Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA535416
Description
This antibody reacts with Bovine Papillomavirus & HPV VLPS types 16 & 11. For IHC applications, it is non-reactive with normal tissue and is non-reactive with cultured cells in immunofluorescent applications. Purity is >90%. This product was purified using a proprietary method resulting in an Ig fraction.
Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
Specifications
| HPV/BPV | |
| Polyclonal | |
| Unconjugated | |
| BPV; HPV; human papillomavirus | |
| Papillomavirus native protein. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG Fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.