Learn More
anti-pan Cadherin, Polyclonal, Abcam™
Brand: Abcam AB6529-200UL
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Chicken, Cow, Dog, Human, Xenopus laevis, Zebrafish. Predicted to work with Rabbit, Bird, Fish, Amphibians
Immunogen: Synthetic peptide: DYDYLNDWGPRFKKLADMYGGGDD conjugated to KLH by a Glutaraldehyde linker, corresponding to C terminal amino acids 889-912 of Chicken pan Cadherin
Applications:
Electron Microscopy, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| pan Cadherin | |
| 64-70mg/mL | |
| Rabbit | |
| 200 μL | |
| Bovine, Canine, Chicken, Fish, Human, Mouse, Rabbit, Rat, Xenopus species, Zebrafish | |
| IgG |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide: DYDYLNDWGPRFKKLADMYGGGDD conjugated to KLH by a Glutaraldehyde linker, corresponding to C terminal amino acids 889-912 of Chicken pan Cadherin | |
| Primary | |
| Antisera |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.