Learn More
peptidyl arginine deiminase, type IV, Mouse, Clone: 4D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PADI4.
Brand: Abnova H00023569-M01.100ug
Description
This gene is a member of a gene family which encodes enzymes responsible for the conversion of arginine residues to citrulline residues. This gene may play a role in granulocyte and macrophage development leading to inflammation and immune response. [provided by RefSeq]
Sequence: AQGTLIRVTPEQPTHAVCVLGTLTQLDICSSAPEDCTSFSINASPGVVVDIAHSPPAKKKSTGSSTWPLDPGVEVTLTMKAASGSTGDQKVQISYYGPKTPPVKALLYLSpecifications
| peptidyl arginine deiminase, type IV | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_012387 | |
| PADI4 | |
| PADI4 (NP_004247, 2 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4D8 | |
| Mouse monoclonal antibody raised against a partial recombinant PADI4. | |
| PADI4 | |
| PAD/PAD4/PADI5/PDI4/PDI5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 23569 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.