Learn More
p62 Ick ligand Mouse anti-Human, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
£157.25 - £561.00
Specifications
| Antigen | p62 Ick ligand |
|---|---|
| Clone | 3 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15392121
|
BD Biosciences
610832 |
50 μg |
£255.00 £157.25 / Each Save £97.75 38% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15805669
|
BD Biosciences
610833 |
150 μg |
£561.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p62 lck ligand (zeta-interacting protein (ZIP)) is a cytoplasmic protein that binds to the SH2 domain of lck (a T cell src tyrosine kinase) in the absence of a phosphotyrosine in either protein. The ubiquitously expressed p62 lck ligand contains a cysteine rich region that is similar to a zinc finger domain, a G protein binding region, a PEST sequence, and several phosphorylation sites. Deletion of the p62 lck ligand N-terminal domain has been reported to abrogate its binding to lck. However, mutation of the tyrosine did not have an effect. In addition, p62 lck ligand binds to the pseudosubstrate region of the PKCζ catalytic domain. In turn, PKCζ phosphorylates p62. p62 lck ligand binds to the dimerization region of PKCζ, thereby inhibiting PKCζ-PKCζ interaction. This suggests that p62 lck ligand may compete with PKCζ. However, it requires PKCζ for proper subcellular localization. These data suggest that p62 lck ligand may be part of the protein bridge that links PKCζ to the tyrosine kinases involved in signaling pathways.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
| p62 Ick ligand | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Zeta Interacting Protein (ZIP); SQSMT1 |
| 3 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human p62 lck ligand aa. 257-437 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.