missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFIT1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA3846

Product Code. 11518851

  • £474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA3-846 detects ISG56 (p56) from mouse samples. PA3-846 has been successfully used in Western blot applications to detect a 39-58 kDa band.

Interferon-induced antiviral RNA-binding protein that specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs and inhibiting expression of viral messenger RNAs. Single-stranded PPP-RNAs, which lack 2'-O-methylation of the 5' cap and bear a 5'-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner. Viruses evolved several ways to evade this restriction system such as encoding their own 2'-O-methylase for their mRNAs or by stealing host cap containing the 2'-O-methylation (cap snatching mechanism). Exhibits antiviral activity against several viruses including human papilloma and hepatitis C viruses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFIT1
Polyclonal
Unconjugated
IFIT1
C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Western Blot
Conc. Not Determined
Whole serum with 0.05% sodium azide
Q64282
IFIT1
aa 217-233 of mouse IFIT1.
RUO
15957
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.