Learn More
p53 Mouse anti-Human, Unlabeled, Clone: DO-7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554294
Description
p53 is a 53kDa nuclear phosphoprotein that acts as a tumor suppressor protein, and is involved in inhibiting cell proliferation when DNA damage occurs. The gene for p53 is the most commonly mutated gene yet identified in human cancers. Missense mutations occur in tumors of the colon, lung, breast, ovary, bladder and several other organs. The mutant p53 is overexpressed in a variety of transformed cells and wildtype p53 forms specific complexes with several viral oncogenes including SV40 large T, E1B from adenovirus, and E6 from human papilloma virus. Wildtype p53 plays a role as a checkpoint protein for DNA damage during the G1/S-phase of the cell cycle. However, it is still unclear, whether point mutated forms of p53 are simple null mutants and/or dominant negatively acting proteins. The DO-7 antibody recognizes human wildtype and mutant p53. It cross-reacts with bovine p53 but does not cross-react with mouse or rat p53. DO-7 recognizes an epitope between amino acids 1-45 of known forms of human p53. Human recombinant p53 protein was used as immunogen. Wildtype 53 proteins have a very short half-life and are usually not detectable with monoclonal antibodies (mAbs) in normal tissues.
Host Species: Mouse
Clone: DO-7
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human Recombinant p53 protein
Formula Weight [Chemical]: 53kDa
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting
Specifications
| p53 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Recombinant p53 protein | |
| 0.1 mg | |
| Cell Biology | |
| Bovine, Human, Monkey | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| DO-7 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.