Learn More
p38 MAPK (pT180/pY182) Mouse, PE-Cy7, Clone: 36/P38 (PT180/PY182), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560241
Description
Activation of the immune and inflammatory responses often involves the recognition of bacterial endotoxin (lipopolysaccharide or LPS). Binding of LPS by monocytes results in the production and release of proinflammatory cytokines, such as IL-1 and TNF. LPS-induced signaling cascades involve members of the Ser/Thr protein kinase family known as the Mitogen Activated Protein Kinases (MAPKs). MAPK signal transduction pathways mediate the effects of various extracellular stimuli on biological processes such as proliferation, differentiation, and death. The p38 MAPKs include p38α (MAPK14), β (MAPK11), γ (MAPK12), and δ (MAPK13). These Ser/Thr kinases are activated by
dual phosphorylation on threonine (T) and tyrosine (Y) within the motif Thr-Gly-Tyr located in kinase subdomain VIII. Activation of p38 MAPK is mediated specifically by the MAP Kinase Kinases, MKK3, MKK4, and MKK6. This leads to the activation of multiple transcription factors (NF-κB, ATF-2, Elk-1, and CHOP) that induce expression of many different genes, including proinflammatory cytokine genes. Thus, p38 MAPKs are central kinases in multiple signal transduction pathways. The 36/p38 (pT180/pY182) monoclonal antibody recognizes the conserved dual phosphorylated site pT180/pY182 of p38α, β, γ, and δ.
Host Species: Mouse
Clone: 36
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human p38 MAPK (pT180/pY182) Peptide
Intracellular Staining
Specifications
| p38 MAPK (pT180/pY182) | |
| Monoclonal | |
| PE-Cyanine7 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Human p38 MAPK (pT180/pY182) Peptide | |
| 50 Tests | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Flow Cytometry | |
| 36/P38 (PT180/PY182) | |
| MAPK14; p38 MAP kinase; p38 MAPK; RK; CSBP2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.