Learn More
purinergic receptor P2X, ligand-gated ion channel, 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human P2RX5 protein.
Brand: Abnova H00005026-D01P.100ug
Description
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Several characteristic motifs of ATP-gated channels are present in its primary structure, but, unlike other members of the purinoceptors family, this receptor has only a single transmembrane domain. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: MGQAGCKGLCLSLFDYKTEKYVIAKNKKVGLLYRLLQASILAYLVVWVFLIKKGYQDVDTSLQSAVITKVKGVAFTNTSDLGQRIWDVADYVIPAQGENVFFVVTNLIVTPNQRQNVCAENEGIPDGACSKDSDCHAGEAVTAGNGVKTGRCLRRENLARGTCEIFAWCPLETSSRPEEPFLKEAEDFTIFIKNHIRFPKFNFSKSNVMDVKDRSFLKSCHFGPKNHYCPIFRLGSVIRWAGSDFQDIALEGGVIGINIEWNCDLDKAASECHPHYSFSRLDNKLSKSVSSGYNFRFARYYRDAAGVEFRTLMKAYGIRFDVMVNGKGAFFCDLVLIYLIKKREFYRDKKYEEVRGLEDSSQEAEDEASGLGLSEQLTSGPGLLGMPEQQELQEPPEAKRGSSSQKGNGSVCPQLLEPHRSTSpecifications
| purinergic receptor P2X, ligand-gated ion channel, 5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human P2RX5 protein. | |
| P2RX5 | |
| MGC47755/P2X5/P2X5R | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5026 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002561.2 | |
| P2RX5 | |
| P2RX5 (NP_002552.2, 1 a.a. ∼ 422 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.