Learn More
p27 (Kip1) Mouse, Unlabeled, Clone: G173-524, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554069
Description
Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catalytic subunits) to form complexes that regulate the progression of the cell cycle. These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small proteins that bind to cyclins, cdks, or cyclin-cdk complexes. These include p15, p16, p18, p19, p21 and p27 [Kip1]. p27 [Kip1] has been shown to inhibit the activity of multiple cyclin-cdk complexes, including cyclin D-cdk4, cyclin E-cdk2 and cyclin A-cdk2. p27 [Kip1] is a 27kDa protein which shares N-terminal sequence homology with p21, and like p21, p27 [Kip1] contains a nuclear localization signal in its C-terminal region. IL-2 activation of T cells has been reported to lead to a decrease in p27 [Kip1] and entry into S phase. Removal of IL-2 from T cell cultures results in increased levels of p27 [Kip1] and cell quiescence.
Host Species: Mouse
Clone: G173-524
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse p27 [Kip1] (full-length) Recombinant Protein
Formula Weight [Chemical]: 27kDa
Bioimaging, Immunoprecipitation, Western Blotting
Specifications
| p27 (Kip1) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse p27 [Kip1] (full-length) Recombinant Protein | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse | |
| IgG1 |
| Western Blot | |
| G173-524 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.