Learn More
p190-B Mouse, Unlabeled, Clone: 54, BD
Mouse Monoclonal Antibody
£343.55
Specifications
| Antigen | p190-B |
|---|---|
| Clone | 54 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15865989
|
BD Biosciences
611613 |
150 μg | |||||||
Description
GTPase activating proteins (GAPs) stimulate the GTP-hydrolyzing activity of GTPases, such as p21ras and Rho. p190-A is a Ras-GAP associated protein that is tyrosine phosphorylated in transformed and growth factor-stimulated cells. Ras-GAP and p190-A are targets of oncoproteins and growth factor receptors. p190-B is another Ras-GAP in the p190 family. It has 51% identity with p190-A and contains several GTPase-related domains in the N-terminal region and a Rho-GAP domain in the C-terminal region. p190-B is expressed in kidney, brain, liver, and lung, as well as in human foreskin fibroblasts, RD muscle cells, and HT-1080 cells. In fibroblasts, p190-B is localized diffusely in the cytoplasm and co-localizes with the α5β1 integrin receptor for fibronectin. Adhesion of fibronectin-coated latex beads to cells leads to the recruitment of p190-B and Rho to the plasma membrane at sites of bead contact. In addition, the recombinant Rho-GAP domain of p190-B displays GAP activity for RhoA, Rac1, and G25K/CDC42Hs. Thus, p190-B is thought to act as a transmembrane link between integrins and Rho GTPases during fibronectin-induced changes in cell morphology and motility.
Host Species: Mouse
Clone: 54
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human p190-B aa. 1102-1214
Immunofluorescence, Western Blotting
Specifications
| p190-B | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 54 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Rat | |
| Human p190-B aa. 1102-1214 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.