missing translation for 'onlineSavingsMsg'
Learn More

p150 [Glued] Mouse, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610473

 View more versions of this product

Product Code. 15895479

  • ONLINE EXCLUSIVE PRICE
    £196.00
    £188.32 / 1 set
    Save £7.68
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

p150 [Glued] was identified as a polypeptide associated with cytoplasmic dynein, the minus-end-directed microtubule-based motor protein. p150 [Glued] is also a member of the oligomeric dynactin complex. Dynactin mediates dynein-driven vesicle motility, as well as lower eukaryote nuclear transport. p150 [Glued] bears significant homology to the product of the Glued gene in Drosophila. It has been shown in vitro to be a required activator of dynein-mediated transport along microtubules. The p150 [Glued] component of the dynactin complex binds to microtubules and the actin-like protein Centractin (Arp-1), another member of the dynactin complex. In the developing rat, p150 [Glued] is expressed at high levels in neural tissue. Microtubule bindings assays with selected constructs of p150 [Glued] indicate that amino acids 39-150 are required for microtubule association.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p150 [Glued]
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
1
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.