Learn More
p130Cas (pY249) Mouse anti-Human, Unlabeled, Clone: J169-757.12.2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558401
Description
p47v-crk (v-Crk) is the product of a transforming gene, v-crk, that was isolated from avian sarcoma viruses. The v-Crk protein is a fusion product of viral Gag protein and a part of cellular Crk that includes SH2 and SH3 domains. v-Crk-induced transformation increases tyrosine phosphorylation of several cellular proteins, including p130Cas (CRK-associated substrate). The p130Cas is tightly associated with v-Crk via the SH2 domain of v-Crk. Tyrosine phosphorylation of p130Cas occurs in conjunction with cellular transformation in cells that express v-Src or v-Crk. This phosphorylation leads to a change in p130Cas localization from the cytoplasm to the cell membrane and, possibly, to the nucleus. Since p130Cas also associates with v-Src, it may be a v-Src substrate. Several phosphorylation sites have been described in p130Cas upon Fibroblast Growth Factor stimulation, and phosphorylated tyrosine (Y249) might function as a binding site for the Crk-adaptor molecule. The J169-757.12.2 monoclonal antibody recognizes the phosphorylated Y249 of human p130Cas. The orthologous phosphorylation sites in mouse and rat p130Cas are Y253 and Y347, respectively.
Western Blotting
Specifications
| p130Cas (pY249) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Phosphorylated Human p130Cas | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2b κ |
| Western Blot | |
| J169-757.12.2 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.