Learn More
p130 [Cas] Mouse, Unlabeled, Clone: 21, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610272
Description
p47 [v-crk] is the product of a transforming gene, v- crk , that was isolated from avian sarcoma viruses. The v-crk protein is a fusion product of the viral gag protein and a part of cellular crk that includes SH2 and SH3 domains. v-crk induced transformation increases tyrosine phosphorylation of several cellular proteins, including p130 [Cas]. p130 [Cas] is tightly associated with v-crk via the SH2 domain of v-crk. Tyrosine phosphorylation of p130 [Cas] occurs in conjunction with cellular transformation in cells that express v-src or v-crk. This phosphorylation leads to a change in p130 [Cas] localization from the cytoplasm to the cell membrane and, possibly, to the nucleus. Since p130 [Cas] also associates with v-src, it may be a v-src substrate.
Specifications
| p130 [Cas] | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 21 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.