Learn More
p130 [Cas] Mouse, Unlabeled, Clone: 21, BD
Description
p47 [v-crk] is the product of a transforming gene, v- crk , that was isolated from avian sarcoma viruses. The v-crk protein is a fusion product of the viral gag protein and a part of cellular crk that includes SH2 and SH3 domains. v-crk induced transformation increases tyrosine phosphorylation of several cellular proteins, including p130 [Cas]. p130 [Cas] is tightly associated with v-crk via the SH2 domain of v-crk. Tyrosine phosphorylation of p130 [Cas] occurs in conjunction with cellular transformation in cells that express v-src or v-crk. This phosphorylation leads to a change in p130 [Cas] localization from the cytoplasm to the cell membrane and, possibly, to the nucleus. Since p130 [Cas] also associates with v-src, it may be a v-src substrate.
Specifications
Specifications
| Antigen | p130 [Cas] |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 21 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.