Learn More
P-glycoprotein Mouse anti-Human, Unlabeled, Clone: 17F9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557001
Description
Reacts with a 170-180kDa transmembrane glycoprotein (P-glycoprotein), a product of the multidrug resistance-1 (MDR1) gene. This glycoprotein is expressed on MDR positive cells and has been reported to be expressed on many normal tissues, such as adrenal glands and endothelium, in the brain and skin. P-glycoprotein is known to impart drug resistance to cells by pumping many anti-cancer drugs out of the cytoplasm. 17F9 antibody binds to the surface of the viable, unfixed MDR+ cell line, PMG-Y (please inquire), and is able to partially block the binding of UIC2 antibody (another MDR-specific monoclonal antibody). Immunoprecipitation application is reported, but not routinely tested in house.
Host Species: Mouse
Clone: 17F9
Isotype: IgG2b, κ
Cross Reactivity: Human
Flow Cytometry, Immunoprecipitation, Western Blotting
Specifications
| P-glycoprotein | |
| Monoclonal | |
| 0.5mg/mL | |
| CD243; ABCB1; ABC20; CLCS; GP170; MDR1; P-GP; PGY1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 17F9 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human MDR1 Transfected Cell Line | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.