missing translation for 'onlineSavingsMsg'
Learn More

OxPhos Blue Native WB Mouse anti-Bovine, Human, Mouse, Invitrogen™

Product Code. 10512144
Change view
Click to view available options
Quantity:
1 Unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10512144 1 Unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10512144 Supplier Invitrogen Supplier No. 457999

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Antibody Cocktail

This Anti-OxPhos Complex Kit is a cocktail of monoclonal antibodies specific to human, bovine, mouse OxPhos Complex This antibody cocktail is validated for use in Western blotting. Mouse monoclonal [20C11B11B11] to (C-I) NDUFA9: Amount: 120 μg Wo

Altered levels of assembly can arise from mutations in individual subunits, mutations in assembly factors for the complex(es), mtDNA depletion or as a result of physiological and or pathological changes e.g. hormone treatment, exercise, diet or oxidative stress. Blue Native polyacrylamide gel electrophoresis (BN-PAGE) is a simple and effective way to subfractionate mitochondrial proteins as intact complexes on a single gel (in one dimension). It can be used to detect altered assembly of these complexes arising from mutations in subunits, mutations in assembly factors, or mtDNA depletion. This type of analysis has been performed with biopsy samples, platelets and fibroblast cells from patients with suspected mitochondrial diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OxPhos Blue Native WB
Applications Western Blot
Classification Cocktail
Clone Cocktail
Concentration 1.5 mg/mL
Conjugate Unconjugated
Gene Alias Oxidative Phosphorylation; OxPhos Complex
Host Species Mouse
Purification Method IgG fraction
Quantity 1 Unit
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Mouse
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.