Learn More
olfactory receptor, family 2, subfamily C, member 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human OR2C1 protein.
Brand: Abnova H00004993-B01.50uL
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Sequence: MDGVNDSSLQGFVLMGISDHPQLEMIFFIAILFSYLLTLLGNSTIILLSRLEARLHTPMYFFLSNLSSLDLAFATSSVPQMLINLWGPGKTISYGGCITQLYVFLWLGATECILLVVMAFDRYVAVCRPLRYTAIMNPQLCWLLAVIAWLGGLGNSVIQSTFTLQLPLCGHRRVEGFLCEVPAMIKLACGDTSLNQAVLNGVCTFFTAVPLSIIVISYCLIAQAVLKIRSAEGRRKAFNTCLSHLLVVFLFYGSASYGYLLPAKNSKQDQGKFISLFYSLVTPMVNPLIYTLRNMEVKGALRRLLGKGREVGSpecifications
| olfactory receptor, family 2, subfamily C, member 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human OR2C1 protein. | |
| OR2C1 | |
| MGC163200/MGC95444/OLFmf3/OR2C2P | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_012368.1 | |
| OR2C1 | |
| OR2C1 (AAI30329.1, 1 a.a. ∼ 312 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 4993 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.