missing translation for 'onlineSavingsMsg'
Learn More
Learn More
opioid receptor, mu 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant OPRM1.
Brand: Abnova H00004988-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MSDAQLGPLRLTLLSVSARTGFCKKQQELWQRRKEAAEALGTRKVSVLLATSHSGARPAVSTMDSSAAPTNASNCTDALAYSSCSPAPSPGSWVNLSHLDGNLSDPCGPNSpecifications
| opioid receptor, mu 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant OPRM1. | |
| OPRM1 | |
| KIAA0403/MOR/MOR1/OPRM | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| 4988 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000914 | |
| OPRM1 | |
| OPRM1 (NP_000905, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction