missing translation for 'onlineSavingsMsg'
Learn More
Learn More
oligodendrocyte transcription factor 1, Mouse, Clone: 2B11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant OLIG1.
Brand: Abnova H00116448-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: PDAKEEQQQQLRRKINSRERKRMQDLNLAMDALREVILPYSAAHCQGAPGRKLSKIATLLLARNYILLLGSSLQELRRALSpecifications
| oligodendrocyte transcription factor 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_138983.1 | |
| OLIG1 | |
| OLIG1 (NP_620450.1, 80 a.a. ∼ 159 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 116448 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 2B11 | |
| Mouse monoclonal antibody raised against a full length recombinant OLIG1. | |
| OLIG1 | |
| BHLHB6/bHLHe21 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction