missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFM1 Monoclonal Antibody (2G12-1B3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521044
This item is not returnable.
View return policy
Description
Peptide Sequence: MPGRWRWQRD MHPARKLLSL LFLILMGTEL TQNKRENKAE KMGGPESERK TTGEKTLNEL PLFCLEAHAG SLALPRMCSP NPNPAVGLCR PAYPQSPSPG AAQTISQSLL ERFCMASRRE VFLAPGRPGG GWWLCTVAGQ MHSFMCTHTH THAHTGEQIP AEKSQPGPD.
The Olfactomedin family comprises a diverse group of secreted glycoproteins, which includes OLFM1 (Noelin-1), OLFM2 (Noelin-2), OLFM3 (Noelin-3), OLFM4 (Noelin-4), tiarin, pancortin, gliomedin and mycocilin. These proteins are implicated in the development of the nervous system. Specifically, OLFM1 and OLFM2 expression is observed in the neural plate and neural crest, as well as in the cranial ganglia in mouse at E8-10, and later in brain tissue and in the zone of polarizing activity in the limb. Overexpression of OLFM1 causes an excess of neural crest emigrations and prolonged neural crest production. OLFM2 participates in the regulation of the development of the anterior nervous system. An Arg144Gln mutation in OLFM2 has been implicated as a possible cause for open-angle glaucoma (OAG).
Specifications
| OLFM1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q99784 | |
| OLFM1 | |
| OLFM1 (AAH00189, 1 a.a. ~ 169 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 2G12-1B3 | |
| Unconjugated | |
| OLFM1 | |
| 1B426B; AMY; AW742568; D2Sut1e; neuroblastoma protein; neuronal olfactomedin-related ER localized protein; Noe1; Noel; Noel1; Noelin; Noelin 1; Noelin 2; NOELIN1; OlfA; olfactomedin 1; olfactomedin related ER localized protein; olfactomedin-1; OLFM1; Pancortin; pancortin 1; Pancortin 1-4; Pancortin3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10439 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction