missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFM2 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA548087
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) Olfactomedin-1 is observed and less than 5% cross-reactivity with recombinant human Olfactomedin-4 and rmOlfactomedin-L3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Involved in transforming growth factor beta (TGF-beta)-induced smooth muscle differentiation. TGF-beta induces expression and nuclear translocation of OLFM2 where it binds to SRF, causing its dissociation from the transcriptional repressor HEY2/HERP1 and facilitating binding of SRF to target genes. Plays a role in AMPAR complex organization.
Specifications
| OLFM2 | |
| Polyclonal | |
| Unconjugated | |
| OLFM2 | |
| A030009A06Rik; neuronal olfactomedin related ER localized protein 2; NOE2; noelin 2; NOELIN2; Noelin-2; NOELIN2_V1; olfactomedin 2; olfactomedin-2; OlfC; OLFM2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 93145 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O95897 | |
| OLFM2 | |
| CHO-derived recombinant human Olfactomedin-2/Noelin-2 Gln21-Pro454. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction