Learn More
anti-ODC1 Matched Antibody Pair, Abnova
Brand: Abnova H00004953-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-ODC1 (100μg); Mouse monoclonal anti-ODC1, IgG2bκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes the rate-limiting enzyme of the polyamine biosynthesis pathway which catalyzes ornithine to putrescine. The activity level for the enzyme varies in response to growth-promoting stimuli and exhibits a high turnover rate in comparison to other mammalian proteins. Originally localized to both chromosomes 2 and 7, the gene encoding this enzyme has been determined to be located on 2p25, with a pseudogene located on 7q31-qter.
ELISA
Specifications
| ODC1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ODC1. | |
| ODC | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ODC1 (100 μg) 2. Detection antibody: mouse monoclonal anti-ODC1, IgG2b Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| ODC1 | |
| ODC1 | |
| Affinity Purified | |
| RUO | |
| 4953 | |
| Human | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.