missing translation for 'onlineSavingsMsg'
Learn More

anti-ODC1 Matched Antibody Pair, Abnova™

Brand:  Abnova H00004953-AP61.1Set

Product Code. 16112394

  • £729.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes: Capture antibody: mouse monoclonal anti-ODC1 (100μg); Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-ODC1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
This item is not returnable. View return policy

Description

Description

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ODC1

Host Species: Mouse, Rabbit
Species Reactivity: Hu

ELISA Pair (Transfected Lysate)

Specifications

Specifications

ODC1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ODC1.
ODC
Mouse, Rabbit
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ODC1 (100 μg) 2. Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-ODC1 (50 μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Unconjugated
ODC1
ODC1
Affinity Purified
RUO
4953
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.