missing translation for 'onlineSavingsMsg'
Learn More

nudix (nucleoside diphosphate linked moiety X)-type motif 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human NUDT2 protein.

Brand:  Abnova H00000318-D01P

Product Code. 16104714

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the MutT family of nucleotide pyrophosphatases, a subset of the larger NUDIX hydrolase family. The gene product possesses a modification of the MutT sequence motif found in certain nucleotide pyrophosphatases. The enzyme asymmetrically hydrolyzes Ap4A to yield AMP and ATP and is responsible for maintaining the intracellular level of the dinucleotide Ap4A, the function of which has yet to be established. This gene may be a candidate tumor suppressor gene. Alternative splicing has been observed at this locus and three transcript variants, all encoding the same protein, have been identified. [provided by RefSeq

Sequence: MALRACGLIIFRRCLIPKVDNNAIEFLLLQASDGIHHWTPPKGHVEPGEDDLETALRETQEEAGIEAGQLTIIEGFKRELNYVARNKPKTVIYWLAEVKDYDVEIRLSHEHQAYRWLGLEEACQLAQFKEMKAALQEGHQFLCSIEA
Specifications

Specifications

nudix (nucleoside diphosphate linked moiety X)-type motif 2
Polyclonal
Rabbit polyclonal antibody raised against a full-length human NUDT2 protein.
NUDT2
APAH1/MGC10404
Rabbit
Affinity Purified
RUO
Primary
Human
Antibody
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_001161
NUDT2
NUDT2 (NP_001152.1, 1 a.a. ∼ 147 a.a) full-length human protein.
100 μg
Apoptosis
318
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.