Learn More
nudix (nucleoside diphosphate linked moiety X)-type motif 1, Mouse, Clone: 5F11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NUDT1.
Brand: Abnova H00004521-M02.100ug
Description
Misincorporation of oxidized nucleoside triphosphates into DNA/RNA during replication and transcription can cause mutations that may result in carcinogenesis or neurodegeneration. The protein encoded by this gene is an enzyme that hydrolyzes oxidized purine nucleoside triphosphates, such as 8-oxo-dGTP, 8-oxo-dATP, 2-hydroxy-dATP, and 2-hydroxy rATP, to monophosphates, thereby preventing misincorporation. The encoded protein is localized mainly in the cytoplasm, with some in the mitochondria, suggesting that it is involved in the sanitization of nucleotide pools both for nuclear and mitochondrial genomes. Several alternatively spliced transcript variants, some of which encode distinct isoforms, have been identified. Additional variants have been observed, but their full-length natures have not been determined. A single-nucleotide polymorphism that results in the production of an additional, longer isoform (p26) has been described. [provided by RefSeq
Sequence: MSGISPQQMGEPEGSWSGKNPGTMGASRLYTLVLVLQPQRVLLGMKKRGFGAGRWNGFGGKVQEGETIEDGARRELQEESGLTVDALHKVGQIVFEFVGEPELMDVHVFCTDSIQGTPVESDEMRPCWFQLDQIPFKDMWPDDSYWFPLLLQKKKFHGYFKFQGQDTILDYTLREVDTVSpecifications
| nudix (nucleoside diphosphate linked moiety X)-type motif 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC014618 | |
| NUDT1 | |
| NUDT1 (AAH14618, 1 a.a. ∼ 179 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Stem Cell Biology | |
| 4521 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 5F11 | |
| Mouse monoclonal antibody raised against a full length recombinant NUDT1. | |
| NUDT1 | |
| MTH1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.