missing translation for 'onlineSavingsMsg'
Learn More

Nucleoporin p62 Mouse, Unlabeled, Clone: 53, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610497

 View more versions of this product

Product Code. 15865489

  • ONLINE EXCLUSIVE PRICE
    £164.00
    £157.25 / 1 set
    Save £6.75
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The nuclear pore complex (NPC) acts as a gate to mediate active transport of proteins and RNA into and out of the nucleus. Proteins actively transported into the nucleus through the NPC require specific nuclear localization sequences. Many of these nucleoporins contain N-acetylglucosamine (GlcNAc) residues that are O-linked to serine or threonine. p62 is the best characterized member of a group of nucleoporins that line the central region of the NPC. A tightly associated complex is formed by p62 and two other nucleoporins, p54 and p58. p54 binds to a carboxy-terminal coiled-coil domain of p62 and p58 binds to a dimer of p54. The amino-terminal domain of p62 contains a series of XFXFX repeats and is joined to the coiled-coil domain by a threonine-rich linker segment. The major role of p62 is maintenance of the structural integrity of NPCs.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nucleoporin p62
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Chicken, Human, Mouse, Rat
IgG2b
Western Blot
53
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.