missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517346
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.Specifications
NSF | |
Polyclonal | |
Unconjugated | |
NSF | |
AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100770898, 18195, 4905, 60355 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
10 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P18708, P46459, P46460, Q9QUL6 | |
NSF | |
Synthetic peptide corresponding to residues surrounding Leu524 of human NSF | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |