missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSDHL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530577
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat. Predicted reactivity: Mouse (91%), Rat (92%), Xenopus laevis (80%), Pig (94%), Rhesus Monkey (97%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.
Specifications
| NSDHL | |
| Polyclonal | |
| Unconjugated | |
| Nsdhl | |
| AI747449; bare patches; Bpa; H105E3; NAD(P) dependent steroid dehydrogenase-like; NAD(P)-dependent steroid dehydrogenase-like protein; Nsdhl; Protein H105e3; SDR31E1; short chain dehydrogenase/reductase family 31E, member 1; sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; Str; striated; XAP104 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50814 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15738 | |
| Nsdhl | |
| Recombinant protein encompassing a sequence within the center region of human NSDHL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction