Learn More
NAD(P) dependent steroid dehydrogenase-like, Mouse, Clone: 6E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NSDHL.
Brand: Abnova H00050814-M01.100ug
Description
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene. [provided by RefSeq]
Sequence: MEPAVSEPMRDQVARTHLTEDTPKVNADIEKVNQNQAKRCTVIGGSGFLGQHMVEQLLARGYAVNVFDIQQGFDNPQVRFFLGDLCSRQDLYPALKGVNTVFHCASPPPSSpecifications
| NAD(P) dependent steroid dehydrogenase-like | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_015922 | |
| NSDHL | |
| NSDHL (NP_057006, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6E3 | |
| Mouse monoclonal antibody raised against a partial recombinant NSDHL. | |
| NSDHL | |
| H105E3/SDR31E1/XAP104 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 50814 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.