missing translation for 'onlineSavingsMsg'
Learn More

anti-NRF2, Clone: EP1808Y, Abcam™

Product Code. 13053475
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
13053475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13053475 Supplier Abcam Supplier No. AB62352100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.

Host Species: Rabbit
Clone: EP1808Y
Isotype: IgG
Cross Reactivity: Human, does not react with Rat
Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human Nrf2

Applications:

Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NRF2
Classification Monoclonal
Clone EP1808Y
Concentration 100μL at mg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues near the C-terminus of human Nrf2
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.