Learn More
NR5A1 Polyclonal Antibody, Invitrogen™
Description
PA1-30599 detects NR5A1 from human samples. PA1-30599 has been successfully used in Western blot applications. The PA1-30599 immunogen is: Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Specifications
| Antigen | NR5A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Gene | NR5A1 |
| Gene Accession No. | Q13285 |
| Gene Alias | AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1 |
| Gene Symbols | NR5A1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.