Learn More
NPC2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NPC2 protein.
Brand: Abnova H00010577-D01P.100ug
Description
This gene encodes a protein containing a lipid recognition domain. The encoded protein may function in regulating the transport of cholesterol through the late endosomal/lysosomal system. Mutations in this gene have been associated with Niemann-Pick disease, type C2 and frontal lobe atrophy. [provided by RefSeq
Sequence: MRFLAATFLLLALSTAAQAEPVQFKDCGSVDGVIKEVNVSPCPTQPCQLSKGQSYSVNVTFTSNIQSKSSKAVVHGILMGVPVPFPIPEPDGCKSGINCPIQKDKTYSYLNKLPVKSEYPSIKLVVEWQLQDDKNQSLFCWEIPVQIVSHLSpecifications
| NPC2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human NPC2 protein. | |
| NPC2 | |
| HE1/MGC1333/NP-C2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10577 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006432.3 | |
| NPC2 | |
| NPC2 (NP_006423.1, 1 a.a. ∼ 151 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Rat, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.