missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP2 Mouse anti-Yeast, Clone: 22G1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110027
This item is not returnable.
View return policy
Description
For Western blot using yeast samples, use a dilution of 1:2000 for cell lysates and 1:10,000 for nuclear fractions.
Nop2 is the yeast homologue of human p120 nucleolar proliferation antigen. This protein is localized in the nucleus.
Specifications
| Nop2 | |
| Monoclonal | |
| Unconjugated | |
| MGC117384, MGC149287, MGC149288, NOL1, NOP120, NSUN1, p120, nop2p | |
| Mouse | |
| 500 μL | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG3 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 22G1 | |
| NOP2 | |
| NOP2 | |
| Yeast nuclear preparation. | |
| RUO | |
| 855664 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction